Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α,2,3,5,6-Heptafluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC3424-5G PC3424-25G
76613-752EA 27 USD InStock
76613-752 76613-750
α,α,α,2,3,5,6-Heptafluorotoluene 98%
α,α,α,2,3,5,6-Heptafluorotoluene


Formula: C₇HF₇
MW: 218.07 g/mol
Boiling Pt: 111…112 °C
Density: 1.601 g/cm³ (25 °C)
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00012385
CAS Number: 651-80-9
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty