Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Difluoromethoxy)benzaldehyde 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC9371-1G PC9371-5G PC9371-10G
76610-332EA 21 USD InStock
76610-332 76610-330 76610-334
3-(Difluoromethoxy)benzaldehyde 99%
3-(Difluoromethoxy)benzaldehyde


Formula: C₈H₆F₂O₂
MW: 172.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236222
CAS Number: 85684-61-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty