Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluoropyridine

Supplier: APOLLO SCIENTIFIC
Danger

PC7262-25G PC7262-100G PC7262-5G
76608-294EA 22 USD InStock
76608-294 76608-292 76608-290
4-Bromo-2-fluoropyridine
4-Bromo-2-fluoropyridine


Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 65 °C/5mmHg
Density: 1.713 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD04112504
CAS Number: 128071-98-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty