Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl trifluoromethanesulfonate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC5170-10G PC5170-100G PC5170-50G
76618-662EA 26 USD InStock
76618-662 76618-660 76618-664
Methyl trifluoromethanesulfonate 98%
Methyl trifluoromethanesulphonate


Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
Boiling Pt: 94…102 °C
Melting Pt: –64 °C
Density: 1.45 g/cm³ (25 °C)
Flash Pt: 38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000409
CAS Number: 333-27-7
EINECS: 206-371-7
UN: 2920
ADR: 8+3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty