Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluorobenzoyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 4-Bromo-2-fluorobenzene-1-carbonyl chloride

PC8531-5G PC8531-25G
76624-544EA 74 USD InStock
76624-544 76624-546
4-Bromo-2-fluorobenzoyl chloride 98%
4-Bromo-2-fluorobenzoyl chloride


Formula: C₇H₃BrClFO
Boiling Pt: 86…88 °C/15mmHg
Melting Pt: 24…25 °C
Storage Temperature: Ambient
MDL Number: MFCD03411584
CAS Number: 151982-51-3
UN: 3261
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty