Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethoxy)benzenesulfonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 4-(Trifluoromethoxy)benzene-1-sulfonyl chloride4-(Trifluoromethoxy)benzene-1-sulphonyl chloride

PC7436R-5G PC7436R-100G PC7436R-25G
76627-454EA 13.39 USD InStock
76627-454 76627-452 76627-450
4-(Trifluoromethoxy)benzenesulfonyl chloride 98%
4-(Trifluoromethoxy)benzenesulphonyl chloride


Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
Boiling Pt: 120…122 °C/15mmHg
Density: 1.536 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042408
CAS Number: 94108-56-2
EINECS: 302-397-9
UN: 3265
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty