Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Fluorophenyl)acetonitrile 90%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Fluorobenzyl cyanide2-(2-Fluorophenyl)acetonitrile

PC5652-500G PC5652-100G
76627-122EA 38 USD InStock
76627-122 76627-120
(2-Fluorophenyl)acetonitrile 90%
(2-Fluorophenyl)acetonitrile


Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 235 °C
Density: 1.059 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00001897
CAS Number: 326-62-5
EINECS: 206-313-0
UN: 3276
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty