Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α,α',α',α'-Hexafluoro-p-xylene 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1,4-Bis(trifluoromethyl)benzeneα,α,α,β,β,β-Hexafluoro-p-xylene

PC3080-500G PC3080-1KG PC3080-100G
76613-702EA 80 USD InStock
76613-702 76613-698 76613-700
α,α,α,α',α',α'-Hexafluoro-p-xylene 98%
α,α,α,α',α',α'-Hexafluoro-p-xylene


Formula: C₈H₄F₆
MW: 214.11 g/mol
Boiling Pt: 115…117 °C
Melting Pt: –1 °C
Density: 1.381 g/cm³ (25 °C)
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00000402
CAS Number: 433-19-2
EINECS: 207-086-0
UN: 1993
ADR: 3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty