Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4-fluoro-1-iodobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 1-Bromo-5-fluoro-2-iodobenzene3-Bromo-1-fluoro-4-iodobenzene2-Bromo-4-fluoroiodobenzene

PC7393-500G PC7393-25G PC7393-100G
76608-434EA 21 USD InStock
76608-434 76608-432 76608-436
2-Bromo-4-fluoro-1-iodobenzene 98%
2-Bromo-4-fluoro-1-iodobenzene


Formula: C₆H₃BrFI
MW: 300.9 g/mol
Boiling Pt: 224…225 °C
Density: 2.323 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00142868
CAS Number: 202865-73-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty