Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(pentafluorophenyl)phosphine 99%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Tris(perfluorophenyl)phosphinePerfluorotriphenylphosphine

PC7905-25G PC7905-5G PC7905-1G
76609-034EA 61 USD InStock
76609-034 76609-030 76609-032
Tris(pentafluorophenyl)phosphine 99%
Tris(pentafluorophenyl)phosphine


Formula: C₁₈F₁₅P
MW: 532.15 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00079654
CAS Number: 1259-35-4
EINECS: 215-021-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty