Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-4-fluorobenzyl alcohol 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC7379-5G PC7379-25G
76608-410EA 13 USD InStock
76608-410 76608-408
3-Bromo-4-fluorobenzyl alcohol 99%
3-Bromo-4-fluorobenzyl alcohol


Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 80 °C/0.5mmHg
Storage Temperature: Ambient
MDL Number: MFCD00143093
CAS Number: 77771-03-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty