Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluorobenzyl alcohol 98+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (4-Bromo-2-fluorophenyl)methanol4-Bromo-2-fluorobenzyl Alcohol

PC7381-25G PC7381-100G PC7381-5G
76608-412EA 94 USD InStock
76608-412 76608-416 76608-414
4-Bromo-2-fluorobenzyl alcohol 98+%
4-Bromo-2-fluorobenzyl alcohol


Formula: C₇H₆BrFO
MW: 205.03 g/mol
Melting Pt: 30…35 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00143265
CAS Number: 188582-62-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty