Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluorosalicylaldehyde

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 5-Fluoro-2-hydroxybenzaldehyde

PC5683-5G PC5683-25G PC5683-100G
76602-648EA 152 USD InStock
76602-648 76602-658 76602-650
5-Fluorosalicylaldehyde
5-Fluorosalicylaldehyde


Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 56 °C/0.75mmHg
Melting Pt: 82…85 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01090997
CAS Number: 347-54-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty