Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-(trifluoromethyl)benzoate 99+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Methyl 3-(trifluoromethyl)benzoate

PC4482-100G PC4482-500G PC4482-25G
76626-512EA 38 USD InStock
76626-512 76626-514 76626-516
Methyl-3-(trifluoromethyl)benzoate 99+%
Methyl 3-(trifluoromethyl)benzoate


Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 80…82 °C/15mmHg
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00043543
CAS Number: 2557-13-3
UN: 3272
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty