Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-3-Fluoropyrrolidine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (S)-3-Fluoropyrrolidine hydrochloride

PC6738-1G PC6738-500MG PC6738-5G
76603-802EA 51 USD InStock
76603-802 76603-804 76603-806
(S)-(+)-3-Fluoropyrrolidine hydrochloride
(S)-(+)-3-Fluoropyrrolidine hydrochloride


Formula: C₄H₉ClFN
MW: 125.57 g/mol
Melting Pt: 183…187 °C
Storage Temperature: Ambient
MDL Number: MFCD04038718
CAS Number: 136725-53-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty