Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Cyano-2-fluorobenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC7963-25G PC7963-500G PC7963-100G
76609-064EA 250 USD InStock
76609-064 76609-060 76609-062
4-Cyano-2-fluorobenzoic acid
4-Cyano-2-fluorobenzoic acid


Formula: C₈H₄FNO₂
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD03094454
CAS Number: 164149-28-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty