Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4-fluorobenzylamine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (2-Bromo-4-fluorophenyl)methanamine hydrochloride

PC7383-5G PC7383-1G PC7383-25G
76608-428EA 114 USD InStock
76608-428 76608-426 76608-424
2-Bromo-4-fluorobenzylamine hydrochloride
2-Bromo-4-fluorobenzylamine hydrochloride


Formula: C₇H₈BrClFN
MW: 240.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672922
CAS Number: 289038-14-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty