Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-2,6-difluoroaniline 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC500008-1G PC500008-250MG
76618-526EA 21 USD InStock
76618-526 76618-524
3-Bromo-2,6-difluoroaniline 95%
3-Bromo-2,6-difluoroaniline


Formula: C₆H₄BrF₂N
MW: 208.006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22627833
CAS Number: 1262198-07-1
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty