Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H,1H,2H,2H-Perfluorooctanesulfonic acid 98%

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 1H,1H,2H,2H-Tridecafluorooctane-1-sulphonic acid

PC6143-100MG PC6143-1G PC6143-250MG
76624-440EA 609 USD InStock
76624-440 76624-442 76624-438
1H,1H,2H,2H-Perfluorooctanesulfonic acid 98%
1H,1H,2H,2H-Perfluorooctanesulfonic acid


Formula: C₈H₅F₁₃O₃S
MW: 428.17 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00042455
CAS Number: 27619-97-2
UN: 3261
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty