Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone 96%

Supplier: APOLLO SCIENTIFIC
Warning

PC502434-5G PC502434-25G PC502434-1G
76606-614EA 169 USD InStock
76606-614 76605-256 76604-530
2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone 96%
2'-Bromo-6'-chloro-2,2,2-trifluoroacetophenone


Formula: C₈H₃BrClF₃O
MW: 287.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062425
CAS Number: 886502-80-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty