Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluoro-6-iodobenzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC52085-250MG PC52085-1G
76626-950EA 177 USD InStock
76626-950 76626-948
4-Bromo-2-fluoro-6-iodobenzonitrile
4-Bromo-2-fluoro-6-iodobenzonitrile


Formula: C₇H₂BrFIN
MW: 325.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28127023
CAS Number: 796968-89-3
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty