Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC535088-100MG PC535088-250MG
76601-548EA 16 USD InStock
76601-548 76601-550
5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one 97%
5-Chloro-4-fluoro-2,3-dihydro-1H-inden-1-one


Formula: C₉H₆ClFO
MW: 184.595 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18208183
CAS Number: 1260013-11-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty