Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trifluoromethylsulfonyl)phenylacetic acid

Supplier: APOLLO SCIENTIFIC
Warning

PC56096-250MG PC56096-1G
76602-252EA 473 USD InStock
76602-252 76602-254
3-(Trifluoromethylsulfonyl)phenylacetic acid
3-(Trifluoromethylsulfonyl)phenylacetic acid


Formula: C₉H₇F₃O₄S
MW: 268.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25460318
CAS Number: 1301739-14-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty