Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-3-hydroxyphenylsulfur pentafluoride

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: 2-Hydroxy-4-(pentafluorothio)aniline

PC5798-1G PC5798-250MG
76603-272EA 527 USD InStock
76603-272 76603-274
4-Amino-3-hydroxyphenylsulfur pentafluoride
4-Amino-3-hydroxyphenylsulfur pentafluoride


Formula: C₆H₆F₅NOS
MW: 235.175 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026459
CAS Number: 1159512-48-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty