Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Difluoromethoxy)-4-fluoroaniline

Supplier: APOLLO SCIENTIFIC
Warning

PC501941-250MG PC501941-5G PC501941-1G
76606-912EA 14 USD InStock
76606-912 76605-526 76607-034
3-(Difluoromethoxy)-4-fluoroaniline
3-(Difluoromethoxy)-4-fluoroaniline


Formula: C₇H₆F₃NO
MW: 177.124 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14698444
CAS Number: 83190-01-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty