Would you like to log in again?
REUZ™ Stainless Steel Beakers
Supplier: United Scientific Supplies
UN3007-3000 UN3007-2000 UN3007-1000 UN3007-500 UN3007-5000 UN3007-100 UN3007-250
470347-980EA 33.15 USD InStock
470347-980 470347-914 470347-910 470347-924 470347-972 470347-986 470347-930
REUZ™ Stainless Steel Beakers
Beakers
Seamless and sanitary stainless steel beakers.
  • 304 stainless steel
  • Reusable and long-lasting
  • Easy to clean
  • Seamless, single piece construction
  • Available in sizes from 100 to 5000 ml
Impact-resistant and built to last, United Scientific REUZ™ ('re-use') beakers are made of 304 stainless steel. The precision corrosion-resistant stainless construction includes rolled top rims, smooth edges and radiused corners for easy, thorough cleaning. These beakers do not contain graduation markings - designed to be filled to the brim to realize full vessel capacity. Useful for storing, mixing, and heating solutions in laboratory and production settings when durability and easy sterilization is desired. Wall thickness is 1 mm.
REUZ™ products promote sustainability in all laboratory environments.
Ordering information: Sold individually.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty