Would you like to log in again?
Union Fittings, Compression, Tee, Jaco Manufacturing®
Supplier: Jaco Manufacturing
70-4-P-PG 200PK 70-6-P-PG 100PK 70-8-P-PG 100PK 70-4-K-PG 100PK 70-2-P-O 200PK
MFLX30805-22BLKEA 414 USD InStock
MFLX30805-22BLK MFLX06382-40BLK MFLX06382-30BLK MFLX06382-05BLK MFLX06382-10BLK
Union Fittings, Compression, Tee, Jaco Manufacturing®
Fittings and Connectors
Excellent choice to connect rigid tubing systems.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Compression fittings offer a more permanent installation. Choose the size and material to suit your application needs.
Good chemical resistance. Usable from -50 to 250 °F (-46 to 121 °C).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty