Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DHR catalyst ≥95%

Supplier: AFG BIOSCIENCE LLC
Synonyms: (2S,3R,4S)-2-Methyltetrahydro-2H-pyran-3,4-diolDihydrorhamnal

C50731-250MG C50731-500MG C50731-1G
76619-238EA 836 USD InStock
76619-238 76619-234 76619-236
DHR catalyst ≥95%
DHR catalyst


Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Storage Temperature: Freezer
CAS Number: 89614-50-6
Order Now
Specification Test Results
Purity Limit > 95%
Appearance Powder or Crystals

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty