Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Essentials Slot Samplers, Sterile, HDPE, Antylia Scientific
Supplier: Antylia Scientific
Essentials
06286-21 06286-22 06286-20 06286-19
76533-708PK 599.75 USD InStock
76533-708 76533-710 76533-712 76533-706
Essentials Slot Samplers, Sterile, HDPE, Antylia Scientific
Samplers Solid Samplers
Reduce cross contamination while sampling various powders, granules, and grains.
  • Suitable for sampling from all layers of the container, providing an accurate representation of contents
  • Constructed of chemically inert high-density polyethylene (HDPE)
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. The samplers are manufactured, assembled, and packed in a Class 100,000 cleanroom to ensure cleanliness. Samplers are also sterilized via gamma irradiation (25 to 45 kGy) and individually bagged to reduce contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty