Would you like to log in again?
MiniVID USB Camera, USB 2.0, 5.1 MP
Supplier: LW SCIENTIFIC SE
MiniVID USB
MVC-U5MP-EMTN
470231-930EA 723.95 USD InStock
470231-930
MiniVID USB Camera, USB 2.0, 5.1 MP
Cameras
The MiniVID USB is our best-selling camera for microscopy.
  • Mac and PC compatible
  • Standard C-mount
  • Standard eyetube (23 mm and 30 mm)
Can be mounted to nearly any brand or type of microscope, either into the eye-tube of a binocular microscope, or thread-mounted onto any trinocular C-mount. The MiniVID USB is the perfect addition to any microscope for training techs, educating clients, or documenting for patient files and medical device records.
Delivery information: Supplied with software, calibration slide, USB cable (1.8 m), and eyepiece adapters for binocular and monocular microscopes (30 and 30.5 mm Ø).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty