Would you like to log in again?
PCR Plates, 96-Well, Semi-Skirted, Roche Style, Azenta Life Sciences
Supplier: Azenta Life Sciences
4TI-0955/0560 4TI-0955
76638-224CS 427.41 USD InStock
76638-224 76638-212
PCR Plates, 96-Well, Semi-Skirted, Roche Style, Azenta Life Sciences
Microplates PCR Plates
Designed for use on Roche LightCycler 480®, 0.1 ml wells, polypropylene, cut corner H12.
  • Subject to a strict QC procedure which includes a visual check and leak testing of every tube
  • Free from DNase, RNase, human genomic DNA, and endotoxin/pyrogen
  • Ultra-smooth, uniform, thin-walled polypropylene wells for optimum PCR and real-time (RT-qPCR) results
  • Low profile 0.1 ml (100 µl) working volume, 0.2 ml (200 µl) total well capacity when used with sealing options
  • Maximum thermal conductivity for efficient heat transfer and precise thermal cycling
  • Raised well rims prevent cross contamination and facilitate effective sealing for reduced evaporation
  • Alphanumeric grid reference to aid well and sample identification
  • Recommended for low volume PCR
  • Compatible with universal standard block thermal cyclers, sequencers and standard multichannel pipettes
This Roche style plate is designed to achieve optimized assay conditions on the Roche LightCycler® 480. This particular style of plate is in a low profile 96-well format, perfect for reaction volumes of 10 to 100 µl.
The wells are shorter than ''standard'' profile wells, which decrease the ''dead space'' between the heated lid of the thermal cycler and the sample in the well. This eliminates condensation forming on the side wall of the tubes, preventing reduction in PCR volume and increasing the efficiency of the reaction. This is especially recommended for reaction volumes below 20 µl.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty