Would you like to log in again?
Anti-IgM Mouse Monoclonal Antibody [clone: R1/69]
Supplier: Diagnostic BioSystems
MOB074 MOB074-RUO MOB074-01 MOB074-05 MOB074-05-RUO MOB074-01-RUO
76633-224EA 316.49 USD InStock
76633-224 CA76633-222 76633-222 CA76633-220 76633-220 CA76633-224
Anti-IgM Mouse Monoclonal Antibody [clone: R1/69]
Antibodies
This antibody reacts with the heavy chain present in all types of human IgM. It stains B cell follicles in human lymphoid tissues. In lymphoid tissues, IgM containing plasma cells stain strongly but immune complexes are usually undetectable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty