Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Anti-CD31 (PECAM-1) Mouse Monoclonal Antibody [clone: JC/70A]
Supplier: Diagnostic BioSystems
MOB034-05 MOB034 MOB034-01-RUO MOB034-RUO MOB034-01 MOB034-05-RUO
76633-126EA 502.8 USD InStock
CA76633-124 76633-126 CA76633-122 76633-124 76633-122 CA76633-126
Anti-CD31 (PECAM-1) Mouse Monoclonal Antibody [clone: JC/70A]
Antibodies
This antibody reacts with a 100 kDa glycoprotein expressed by endothelial cells and 130 kDa glycoprotein present in platelets. It stains endothelial cells in normal as well as malignant tissues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty