Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Anti-CA125 Mouse Monoclonal Antibody [clone: OV185:1]
Supplier: Diagnostic BioSystems
MOB110-01-RUO MOB110 MOB110-05 MOB110-05-RUO MOB110-01 MOB110-RUO
76633-308EA 521.91 USD InStock
CA76633-308 CA76633-306 76633-308 CA76633-304 76633-306 76633-304
Anti-CA125 Mouse Monoclonal Antibody [clone: OV185:1]
Antibodies
This antibody reacts specifically with CA 125 ovarian cancer antigen. Several studies have shown that CA 125 is a useful tumor marker for ovarian epithelial malignancies. However, CA 125 has been described in other neoplasms such as seminal vesicle and anaplastic lymphomas.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty