Would you like to log in again?
Anti-PSMA Mouse Monoclonal Antibody [clone: 3E6]
Supplier: Diagnostic BioSystems
MOB440-01 MOB440 MOB440-05 MOB440-RUO MOB440-05-RUO MOB440-01-RUO
76633-918EA 1028.68 USD InStock
CA76633-918 76633-918 CA76633-916 76633-916 CA76633-920 76633-920
Anti-PSMA Mouse Monoclonal Antibody [clone: 3E6]
Antibodies
PSMA is a 750 amino acid type II membrane glycoprotein. PSMA is expressed in normal and malignant prostatic epithelium and in a subset of non-prostatic tissues. PSMA expression has been shown to correlate with the progression of prostate cancer, with highest levels expressed in hormone-refractory and metastatic disease. PSMA expresses in the neovasculature of a variety of non-prostatic solid tumors.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty