Would you like to log in again?
Powerblanket® Hot Boxes
Supplier: POWERBLANKET LLC MS
Powerblanket®
HB54-1200 HB48-1200
104018-908EA 2118.15 USD InStock
104018-908 104018-956
Powerblanket® Hot Boxes
Controllers Temperature Controllers
Powerbanket HB54 model 48×40×48 inch bulk material warmers offer cutting-edge technology for freeze protection, chemical storage, and viscosity control in a flexible, easy-to-install design that will effectively, evenly, and reliably heat your materials. Our patented design eliminates hot and cold spots by applying uniform full-coverage heat across the entire surface of the container, allowing users to keep materials at the proper viscosity while remaining at specified thermal levels.
  • Powerblanket - designed for any definitive heating applications, surrounded by a rugged vinyl shell in a wraparound design with a removable top, safely raises material temperatures as high as 145 °F and can be used in temperatures as low as −20 °F
  • Powerblanket Xtreme (Roofer’s Hot Box)- designed for harsh climates, surrounded by a rugged vinyl shell in a wraparound design with a removable top, safely raises material temperatures as high as 145 °F and can be used in temperatures as low as −20 °F
  • Maintains an internal temperature as high as of 120 °F / 49 °C (± 5 °F / 3 °C)
  • Provides an insulated full-wrap design
  • Delivers uniform heat to temperature sensitive materials & products
  • Offers quick and easy assembly for portability and storage
  • Localize heat and save money by not heating a warehouse or building
  • Preserve expensive materials without scorching or burning
  • Prevent waste by maintaining consistent temperatures
  • Is a great solution for palletized products: paint, caulk, resins and epoxies, etc.
  • Features top and side access doors
  • 6 foot power cord
  • Custom sizes available upon request
Certifications: Certified by ETL to UL & CSA safety standards
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty