Would you like to log in again?
Bucket Liners, Sterile, STX7099, Texwipe
Supplier: Texwipe
STX7099
10127-114CS 338.56 USD InStock
10127-114
Bucket Liners, Sterile, STX7099, Texwipe
Bags Waste Disposal Bags
Irradiated bucket liners for use with round buckets.
Not recommended for use with TX7054 or TX7063.
29.5" x 45" x 4 mil thick (74.9 cm x 114.2 cm x 0.1 mm thick)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty