Would you like to log in again?
Anti-Cyclin D1 Rabbit Monoclonal Antibody [clone: SP4]
Supplier: Diagnostic BioSystems
RMAB003-RUO RMAB003-05-RUO RMAB003 RMAB003-05 RMAB003-01 RMAB003-01-RUO
76634-706EA 1027.09 USD InStock
CA76634-704 76634-706 CA76634-706 76634-702 76634-704 CA76634-702
Anti-Cyclin D1 Rabbit Monoclonal Antibody [clone: SP4]
Antibodies
Cyclin D1 belongs to the Cyclin D family. Cyclin D1 is required for the cell cycle G1/S transition. Amplification or overexpression of Cyclin D1 plays a pivotal role in the development of various human cancers including breast cancer, colon cancer, melanoma, prostate cancer and lymphoma. It is useful to differentiate mantle cell lymphoma from small cleaved cell lymphoma. Rabbit monoclonal antibodies to Cyclin D1 showed the highest sensitivity to detect this antigen in formalin fixed paraffin embedded tissue as compared to several other clones.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty