Would you like to log in again?
Anti-Alpha-1-antichymotrpsin Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP047 RP047-01 RP047-05 RP047-RUO RP047-01-RUO RP047-05-RUO
76635-042EA 105.1 USD InStock
76635-042 76635-046 CA76635-044 76635-044 CA76635-042 CA76635-046
Anti-Alpha-1-antichymotrpsin Rabbit Polyclonal Antibody
Antibodies
The antibody is specific against human α-1-antichymotrypsin. This antibody stains monocytes and macrophages in human tissues and does not stain any other cell type.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty