Would you like to log in again?
Anti-TTF-1 Mouse Monoclonal Antibody [clone: 8G7G3/1]
Supplier: Diagnostic BioSystems
PDM104-RUO PDM104
76634-352EA 391.73 USD InStock
CA76634-352 76634-352
Anti-TTF-1 Mouse Monoclonal Antibody [clone: 8G7G3/1]
Antibodies
This antibody is specific to a 40 kDa protein, which is identified as thyroid transcription factor (TTF-1). TTF-1 is expressed in epithelial cells of the thyroid gland and lung. TTF-1 stains primary lung adenocarcinoma and small cell carcinoma but does not stain colon and breast carcinoma.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty