Would you like to log in again?
Anti-Inhibin Mouse Monoclonal Antibody [clone: R1]
Supplier: Diagnostic BioSystems
PDM178-RUO PDM178
76634-454EA 359.48 USD InStock
76634-454 CA76634-454
Anti-Inhibin Mouse Monoclonal Antibody [clone: R1]
Antibodies
Inhibin is a dimeric glycoprotein hormone from the TGF-β family made of α and β subunits. This antibody recognizes the 32kDa α subunit of human inhibin. Inhibin α is expressed in a wide variety of human tissues, such as brain, prostate, adrenal, granulose cells of ovary, sertoli cells of testis and in fetoplacental unit.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty