Would you like to log in again?
Anti-CD138 Rabbit Monoclonal Antibody [clone: EP201]
Supplier: Diagnostic BioSystems
RMAB040-05 RMAB040-01 RMAB040 RMAB040-RUO RMAB040-01-RUO RMAB040-05-RUO
76634-782EA 856.31 USD InStock
76634-782 CA76634-780 CA76634-782 76634-780 CA76634-778 76634-778
Anti-CD138 Rabbit Monoclonal Antibody [clone: EP201]
Antibodies
CD138, also known as Syndecan-1, is a member of the transmembrane heparan sulfate proteoglycan family, acts as an extracellular matrix receptor and is involved in many cellular functions, including cell-cell adhesion and cell-matrix adhesion. CD138 expression is found in both hematopoietic and non-hematopoietic cells. In the hematopoietic system, CD138 labels plasma cells. It is an excellent marker for plasmacytic differentiation within the spectrum of hematologic malignancy. Among non-hematolymphoid cells, CD138 reactivity is observed in many types of epithelial cells and stoma cells in both normal and tumor tissues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty