Would you like to log in again?
Anti-CD8 T cell Rabbit Monoclonal Antibody [clone: SP16]
Supplier: Diagnostic BioSystems
RMAB012-01-RUO RMAB012-05 RMAB012-RUO RMAB012-01 RMAB012 RMAB012-05-RUO
76634-740EA 702.24 USD InStock
76634-740 CA76634-738 76634-738 76634-742 CA76634-740 CA76634-742
Anti-CD8 T cell Rabbit Monoclonal Antibody [clone: SP16]
Antibodies
This antibody recognizes the CD8. The CD8 molecule consists of two chains, α and β, which are expressed as a disulphide-linked α/β heterodimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterodimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not β chain).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty