Would you like to log in again?
Anti-Flt-1 Rabbit Polyclonal Antibody
Supplier: Diagnostic BioSystems
RP077-05-RUO RP077-01 RP077-01-RUO RP077-RUO RP077-05 RP077
76635-098EA 546.99 USD InStock
CA76635-096 76635-098 76635-096 CA76635-100 CA76635-098 76635-100
Anti-Flt-1 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 180 kDa protein, known as Flt-1 or vascular endothelial growth factor receptor 1 (VEGFR1). Flt-1 is a cell membrane receptor kinase. Flk-1 has a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty