Would you like to log in again?
AimStrip® Complete Urinalysis System Promo, Germaine Laboratories
Supplier: Germaine Laboratories
AimStrip®
UAPROMO23
76635-422EA 1347.14 USD InStock
76635-422
AimStrip® Complete Urinalysis System Promo, Germaine Laboratories
Clinical Diagnostic Supplies Clinical Analysers
The AimStrip® Urinalysis system is for the semi-quantitative detection of the following analytes in urine: Glucose, Bilirubin, Ketone (Acetoacetic acid), Specific Gravity, Blood, pH, Protein, Urobilinogen, and Leukocytes as well as the qualitative detection of Nitrite.
  • CLIA Waived!
  • Read all combinations of strips
  • Automatic calibration for accurate results
  • Continuous Test Mode performs up to 120 tests per hour. Single Test Mode performs 60 tests / hour.
  • Automatically saves the last 2000 results
  • Includes RS323C and USB ports to export results
  • Can transfer to LIS System or suitable PC software
  • Program languages: English, French, Spanish, Italian and Portuguese
Can be used for general evaluation of health, and aids in the diagnosis and monitoring of metabolic or systemic diseases that affect kidney function, endocrine disorders and diseases or disorders of the urinary tract.
Promo Includes an AimStrip Urine Analzyer II, 4 bottles of 10-SG Test Strips, External QC material, Printer Paper.
Certifications: CLIA Waived
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty