Would you like to log in again?
Whatman™ HEPA-Cap Vent Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ HEPA-CAP™
2809T 111721
76635-532PK 424.9 USD InStock
CA76635-531 76635-532
Whatman™ HEPA-Cap Vent Filters, Whatman products (Cytiva)
Filters Venting Filters
Whatman HEPA-CAP Vent Filters are high-retention and high flow-rate glass fiber filters that are commonly used in both research and industry to remove particulates from air and gases.
  • Typical retention efficiency: 99.97% of all particles ≥ 0.3 µm in air
  • Depth filter design: allows for high loading capacity and bi-directional flow
  • Repeatedly autoclavable: at 121 °C for 20 minutes (132 °C max)
Whatman HEPA-CAP Vent Filters are used across scientific, research, and industrial environments in a variety of air and gas filtration applications that require high particle retention and the ability to capture dirt and debris at high flow rates.
In-line filter capsules are particularly suitable for sterile venting of fermenters or reaction vessels to prevent airborne particles from entering or exiting the vessel. They are also well suited for tissue culture, in-line filtration, and prefiltration for suction or to serve gas in-line filters. Filter media- glass microfiber (GMF).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty