Would you like to log in again?
Eppendorf Tubes® BioBased Tubes, Sterile
Supplier: Eppendorf
Eppendorf Tubes®
0030122526 0030122534 0030122526 0030122518 0030122534 0030122518 0030122542 0030122542
76587-088PK 106.7 USD InStock
76587-088 CA76587-088 76587-090 CA76587-090 76587-092 CA76587-092 76587-094 CA76587-094
Eppendorf Tubes® BioBased Tubes, Sterile
Tubes Centrifuge Tubes
Eppendorf Tubes BioBased are made with 90% biobased plastic. The plastic can be traced back to biological waste material which is attributed to these tubes via the ISCC mass balance approach. By choosing these tubes, customers help meet sustainability goals in their laboratories.
  • Cap with flattened and grooved sides provides user-friendly, ergonomic handling
  • Stable upright positioning with minimal contamination risk
  • Reliable and safe labelling due to large writing area on tube and flat and light-colored cap
  • High g-Safe centrifugation stability
  • Precise dimensions enable maximum compatibility with centrifuge rotors, mixers, and shakers
  • Optimal sample and pellet visibility
  • Pyrogen-, DNase-, RNase-, human and bacterial DNA-free
The Eppendorf tubes biobased in the sizes 5, 15, 25, 50 ml open up doors to make laboratory work significantly more sustainable without jeopardizing experimental results. By using a second-generation biobased polymer made from 90% renewable resources (e.g. waste and residues from vegetable oil refining or waste cooking oil) the product-related carbon footprint of these tubes was significantly reduced.
'General Quality Certificates' on compliance with standards and general quality requirements, product-specific and lot-specific purity certificates as well as 'ACT Certificates' (the independent environmental impact factor certification by my green lab) certify the well-known premium product quality you can expect from Eppendorf tubes combined with a sustainability factor that will clearly be visible in your laboratory carbon footprint with continuous use of these tubes. Like our tubes made of virgin polypropylene, these tubes are also your solution for cell culture and cell biology applications as well as for sample preparation protocols in microbiology and molecular biology laboratories.
Certifications: ISCC PLUS, ACT, My Green Lab Certificates, Premium USP Class 6.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty