Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GeneMate IsoFreeze™ MCT Racks, Scientific Specialties

Supplier: Scientific Specialties
5630-T4-15IS1
490004-208CS 757.55 USD InStock
490004-208
GeneMate IsoFreeze™ MCT Racks, Scientific Specialties
Racks Tube Racks
IsoFreeze MCT racks are manufactured with a gel-filled center section that uses temperature-dependent color-change material, green to yellow or purple to pink.

  • Maintain a sample temperature of less than 4 °C for up to 4 hours when used at room temperature
  • Color change material enables you to see how cold your samples are at a glance
  • IsoFreeze color change racks undergo a dramatic change to a contrasting color at 7 °C

Tube capacity: 24× 1.5 or 2.0 ml snap or screw-cap style tubes.

Ordering information: Racks come with lid.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty