Would you like to log in again?
PCR Foil Adhesive Seal, Peelable, Pierceable, Strong Adhesive
Supplier: Azenta Life Sciences
4TI-0500FL
76637-706CS 170.84 USD InStock
76637-706
PCR Foil Adhesive Seal, Peelable, Pierceable, Strong Adhesive
Seals Microplate Seals
The PCR aluminum foil seal has a strong acrylic adhesive which produces a seal of high integrity.
  • Pierceable
  • Peelable
  • Perforated end tabs for easy application and removal by peeling
  • Temperature range: –80 to +110 °C
  • Non sterile
Adhesive seals provide a versatile, yet robust, method for protecting samples.
To obtain the best sealing results with adhesive seals, we strongly recommend the use of high quality plates with raised rim sealing rings for optimal sealing integrity and guaranteed flatness. All Azenta PCR plates are designed with these features.
Accessories information: Adhesive seal roller; ensures consistent seal application across all wells, 1 roller.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty