Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tube and Cuvette Racks, Eppendorf
Supplier: Eppendorf
0030119860
76338-494EA 28.85 USD InStock
76338-494
Tube and Cuvette Racks, Eppendorf
Racks Tube Racks
Made from high quality Polypropylene. Benchtop racks are essential laboratory accessories for efficient processing, transport, and short-term storage of sample preparation tubes, cryogenic tubes, and cuvettes.
  • Space-saving, all racks are stackable to save lab space when not in use
  • Safe, non-slip silicone feet and a tilt-proof footprint design ensure safe handling
  • Compact dimensions require minimal space
  • Temperature resistance from −86 to +121° C
  • Very good chemical resistance
The comprehensive tube rack and cuvette rack system supports a clearly arranged and error-free handling of reaction vessels in manifold laboratory workflows. Optimized dimensions, design, and functionality result in various user benefits such as easy tube accessibility or flexible usage. The racks offer the optimal solution and perfect fit for all tubes and cuvettes commonly used in laboratories.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty